3-Fluor-5-(trifluormethoxy)benzylamin 97%

Lieferant: Thermo Scientific
Gefahr

H34187.03EA 118 EUR
H34187.03 H34187.MD
3-Fluor-5-(trifluormethoxy)benzylamin 97%
3-Fluor-5-(trifluormethoxy)benzylamin
Formel: C₈H₇F₄NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18783229
CAS-Nummer: 1353000-19-7
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR