2-Fluor-4-methoxybenzylamin

Lieferant: CHEMPUR

CHMPFL13339.POREA 0 EUR
CHMPFL13339.POR
2-Fluor-4-methoxybenzylamin
2-Fluor-4-methoxybenzylamin
Formel: C₈H₁₀FNO
Molecular Weight: 155,1700 g/mol
MDL: MFCD04116358
CAS-Nummer: 937843-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR