4-Fluor-3-methylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL483567-25GEA 221 EUR
SIAL483567-25G SIAL483567-5G
4-Fluor-3-methylphenylboronsäure, Sigma-Aldrich®
4-Fluor-3-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95
MDL: MFCD01863527
CAS-Nummer: 139911-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR