4-Fluor-2-methylphenylboronsäure

Lieferant: CHEMPUR

CHMPFL13844.POREA 0 EUR
CHMPFL13844.POR
4-Fluor-2-methylphenylboronsäure
4-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95
MDL: MFCD02093072
CAS-Nummer: 139911-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR