7-Amino-2-methylchromon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL578231-1GEA 33.1 EUR
SIAL578231-1G
7-Amino-2-methylchromon, Sigma-Aldrich®
7-Amino-2-methylchromon
Formel: C₁₀H₉NO₂
MDL: MFCD04974088
CAS-Nummer: 30779-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR