6-Methylchromon Hydrat 99%

Lieferant: Thermo Scientific

ACRO373020050EA 72.5 EUR
ACRO373020050
6-Methylchromon Hydrat 99%
6-Methylchromon Hydrat
Formel: C₁₀H₈O₂·xH₂O
Molecular Weight: 160,17 g/mol (anhy)
Schmelzpunkt: 76…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00209598
CAS-Nummer: 207511-19-1

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 75 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR