6-Methylchromon

Lieferant: CHEMPUR

CHMPFL12047.POREA 0 EUR
CHMPFL12047.POR
6-Methylchromon
6-Methylchromon
Formel: C₁₀H₈O₂
Molecular Weight: 160,17 g/mol
MDL: MFCD00218598
CAS-Nummer: 38445-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR