2,3-Dichlor-6-fluorbenzylamin 97%

Lieferant: Thermo Scientific
Gefahr

H26424.03EA 113 EUR
H26424.03 H26424.MD
2,3-Dichlor-6-fluorbenzylamin 97%
2,3-Dichlor-6-fluorbenzylamin
Formel: C₇H₆Cl₂FN
Lagertemperatur: Raumtemperatur
MDL: MFCD06660174
CAS-Nummer: 886501-27-5
EINECS: None
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR