2-Bromethyl-phenylsulfid

Lieferant: CHEMPUR

CHMPFL3535.POREA 0 EUR
CHMPFL3535.POR
2-Bromethyl-phenylsulfid
2-Bromethyl-phenylsulfid
Formel: C₈H₉BrS
Molecular Weight: 217,12
MDL: MFCD00040826
CAS-Nummer: 4837-01-8
EINECS: 225-422-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR