4-(Nonyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-9039-1GEA 393 EUR
COBBBB-9039-1G
4-(Nonyloxy)phenylboronsäure
4-(Nonyloxy)phenylboronsäure
Formel: C₁₅H₂₅BO₃
Molecular Weight: 264,1700 g/mol
MDL: MFCD04039033
CAS-Nummer: 173392-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR