4-(Hexyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-2734-1GEA 43.5 EUR
COBBBB-2734-1G COBBBB-2734-25G COBBBB-2734-5G
4-(Hexyloxy)phenylboronsäure
4-(Hexyloxy)phenylboronsäure
Formel: C₁₂H₁₉BO₃
Molecular Weight: 222,09
MDL: MFCD04039027
CAS-Nummer: 121219-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR