3-Isopropoxyphenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL542458-10GEA 316 EUR
SIAL542458-10G SIAL542458-1G
3-Isopropoxyphenylboronsäure, Sigma-Aldrich®
3-Isopropoxyphenylboronsäure
Formel: C₉H₁₃BO₃
Molecular Weight: 180,01
MDL: MFCD03701512
CAS-Nummer: 216485-86-8