2,6-Diisopropylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL719714-1GEA 131 EUR
SIAL719714-1G
2,6-Diisopropylphenylboronsäure, Sigma-Aldrich®
2,6-Diisopropylphenylboronsäure
Formel: C₁₂H₁₉BO₂
MDL: MFCD16876786
CAS-Nummer: 363166-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR