4-Pentylphenylboronsäure ≥97%

Lieferant: Thermo Scientific

Achtung

L18011.06EA 431 EUR
L18011.06 L18011.MD L18011.03
4-Pentylphenylboronsäure ≥97%
4-Pentylphenylboronsäure
Formel: C₁₁H₁₇BO₂
Molecular Weight: 192,07
Schmelzpunkt: 82…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00995151
CAS-Nummer: 121219-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR