4-Hexylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5656-5GEA 323 EUR
COBBBB-5656-5G COBBBB-5656-1G
4-Hexylphenylboronsäure
4-Hexylphenylboronsäure
Formel: C₁₂H₁₉BO₂
Molecular Weight: 206,09 g/mol
MDL: MFCD04039029
CAS-Nummer: 105365-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR