4-Heptylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5866-5GEA 1140 EUR
COBBBB-5866-5G COBBBB-5866-1G
4-Heptylphenylboronsäure
4-Heptylphenylboronsäure
Formel: C₁₃H₂₁BO₂
MDL: MFCD04039028
CAS-Nummer: 256383-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR