4-tert-Butylphenylboronsäure

Lieferant: CHEMPUR

CHMPFL10982.POREA 0 EUR
CHMPFL10982.POR FLUO010982.25G FLUO010982.5G
4-tert-Butylphenylboronsäure
4-tert-Butylphenylboronsäure
Formel: C₁₀H₁₅BO₂
Molecular Weight: 178,04 g/mol
MDL: MFCD01009697
CAS-Nummer: 123324-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR