3-tert-Butylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBFA-1936-5GEA 191 EUR
COBBFA-1936-5G
3-tert-Butylphenylboronsäure
3-tert-Butylphenylboronsäure
Formel: C₁₀H₁₅BO₂
Molecular Weight: 178,0400 g/mol
MDL: MFCD11110647
CAS-Nummer: 560132-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR