Natrium-1-tetradecansulfonat, Supelco®

Lieferant: Merck


M-Clarity™ quality level = MQ100

87191-10GEA 155 EUR
87191-10G
Natrium-1-tetradecansulfonat, Supelco®
Natrium-1-tetradecansulfonat
Formel: C₁₄H₂₉NaO₃S
MDL: MFCD00043415
CAS-Nummer: 6994-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR