1-Nonansulfonsäure Natriumsalz

Lieferant: Molekula

MOLE44958889-100GEA 774 EUR
MOLE44958889-100G
1-Nonansulfonsäure Natriumsalz
1-Nonanesulphonic acid sodium salt
Formel: C₉H₁₉NaO₃S
Molecular Weight: 230,30
MDL: MFCD00025010
CAS-Nummer: 35192-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR