3-Cyan-6,7-dimethylchromon

Lieferant: CHEMPUR

CHMPFL12024.POREA 0 EUR
CHMPFL12024.POR
3-Cyan-6,7-dimethylchromon
3-Cyan-6,7-dimethylchromon
Formel: C₁₂H₉NO₂
CAS-Nummer: 94978-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR