4-Aminotetrahydropyran, Sigma-Aldrich®

Lieferant: Merck

SIAL711357-1GEA 38.5 EUR
SIAL711357-1G
4-Aminotetrahydropyran, Sigma-Aldrich®
4-Aminotetrahydropyran
Formel: C₅H₁₁NO
Molecular Weight: 101,1469 g/mol
MDL: MFCD02179436
CAS-Nummer: 38041-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR