4-Aminotetrahydropyran

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO023514EA 0 EUR
FLUO023514 FLUO023514.5G FLUO023514.1G
4-Aminotetrahydropyran
4-Aminotetrahydropyran
Formel: C₅H₁₁NO
Molecular Weight: 101,1469 g/mol
MDL: MFCD02179436
CAS-Nummer: 38041-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR