Tantaldiborid, Sigma-Aldrich®

Lieferant: Merck

SIAL336173-5GEA 79.1 EUR
SIAL336173-5G
Tantaldiborid, Sigma-Aldrich®
Tantalum diboride
Formel: B₂Ta
CAS-Nummer: 12007-35-1
EINECS: 234-507-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR