Tantalborid, Sigma-Aldrich®

Lieferant: Merck

SIAL336165-5GEA 34.4 EUR
SIAL336165-5G
Tantalborid, Sigma-Aldrich®
Tantalum boride
Formel: BTa
MDL: MFCD00049565
CAS-Nummer: 12007-07-7
EINECS: 234-496-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR