Silicon nitride
Supplier: LGC Standards PROMOCHEM
Synonyms:
Silicon(IV) nitride
PROONIST-656EA
825
EUR
PROONIST-656
Silicon nitride
Silicon nitride
Formula:
N₄Si₃ MW: 140,28 g/mol |
MDL Number:
MFCD00011230 CAS Number: 12033-89-5 EINECS: 234-796-8 |