Kaliumhexafluoronickelat(IV), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL390011-10GEA
128
EUR
SIAL390011-10G
Kaliumhexafluoronickelat(IV), Sigma-Aldrich®
Potassium hexafluoronickelate(IV)
Formel:
F₆K₂Ni Molecular Weight: 250,88 g/mol |
MDL:
MFCD00083064 CAS-Nummer: 17218-47-2 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...