2-Fluoro-5-(4-methoxybenzylcarbamoyl)benzeneboronic acid, 97%

H54400.03EA 257 EUR
ALFAH54400.03 ALFAH54400.MD
2-Fluoro-5-(4-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO4
Formula Weight: 303.10
MDL No.: MFCD20265261
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR