2-Chlor-5-(3-fluorbenzylcarbamoyl)phenylboronsäure ≥97%

Achtung

H55931.03EA 675 EUR
H55931.03 H55931.MD
2-Chlor-5-(3-fluorbenzylcarbamoyl)phenylboronsäure ≥97%
2-Chlor-5-(3-fluorbenzylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₂BClFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20040333
CAS-Nummer: 1449133-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR