2-Chloro-5-(4-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
H55942.MDEA 76.5 EUR
ALFAH55942.MD ALFAH55942.03
2-Chloro-5-(4-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO4
Formula Weight: 319.55