5-Benzylcarbamoyl-2-chlorobenzeneboronic acid, 97%

H54937.03EA 219 EUR
ALFAH54937.03 ALFAH54937.MD
5-Benzylcarbamoyl-2-chlorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H13BClNO3
Formula Weight: 289.52
MDL No.: MFCD22125102
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR