3-Chloro-4-(3-methylbenzylcarbamoyl)benzeneboronic acid, 97%

H54584.03EA 170 EUR
ALFAH54584.03 ALFAH54584.MD
3-Chloro-4-(3-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD20265296
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR