3-Chlor-4-ethoxy-5-fluorbenzylamin 97%, JRD
|
Gefahr
|
H31681.MDEA
55.1
EUR
H31681.MD
H31681.03
3-Chlor-4-ethoxy-5-fluorbenzylamin 97%, JRD
3-Chlor-4-ethoxy-5-fluorbenzylamin
Formel:
C₉H₁₁ClFNO Molecular Weight: 203,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832346 CAS-Nummer: 1017779-02-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...