2-(Methylthio)benzylamin

Lieferant: Thermo Scientific

MAYBKM09736DAEA 0 EUR
MAYBKM09736DA MAYBKM09736CB MAYB04-9736
2-(Methylthio)benzylamin
2-(Methylthio)benzylamin
Formel: C₈H₁₁NS
MDL: MFCD00831039
CAS-Nummer: 56004-83-2
EINECS: 000-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR