4-Hydroxy-3-methoxybenzylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Aminomethyl)-2-methoxyphenol hydrochloride

M-Clarity™ quality level = MQ200

H36605-1G
SIALH36605-1GEA 35.7 EUR
SIALH36605-1G
4-Hydroxy-3-methoxybenzylamin Hydrochlorid, Sigma-Aldrich®
4-Hydroxy-3-methoxybenzylamin Hydrochlorid
Formel: C₈H₁₂ClNO₂
Molecular Weight: 189,64 g/mol
MDL: MFCD00012864
CAS-Nummer: 7149-10-2
EINECS: 230-468-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR