3'-Iod-4'-methoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL631515-5GEA 66.3 EUR
SIAL631515-5G
3'-Iod-4'-methoxyacetophenon, Sigma-Aldrich®
3'-Iod-4'-methoxyacetophenon
Formel: C₉H₉IO₂
Molecular Weight: 276,07 g/mol
MDL: MFCD02258873
CAS-Nummer: 79324-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR