3-Methoxy-2-(trifluormethyl)benzamid ≥97%
|
Achtung
|
H33368.03EA
182
EUR
H33368.03
H33368.MD
3-Methoxy-2-(trifluormethyl)benzamid ≥97%
3-Methoxy-2-(trifluormethyl)benzamid
Formel:
C₉H₈F₃NO₂ Molecular Weight: 219,16 g/mol Schmelzpunkt: 103…205 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185678 CAS-Nummer: 1214351-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...