3-(Methylthio)-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5013-1GEA 786 EUR
COBBBB-5013-1G
3-(Methylthio)-5-(trifluormethyl)phenylboronsäure
3-(Methylthio)-5-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₂S
MDL: MFCD09800876
CAS-Nummer: 957120-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR