2-Hydroxy-4-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS INC.
COBBBB-5333-1GEA
507
EUR
COBBBB-5333-1G
2-Hydroxy-4-(trifluormethyl)phenylboronsäure
2-Hydroxy-4-(trifluormethyl)phenylboronsäure
Formel:
C₇H₆BF₃O₃ Molecular Weight: 205,9270 g/mol |
MDL:
MFCD03094996 CAS-Nummer: 1072951-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...