2-Cyan-4-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-8820-1GEA 140 EUR
COBBPN-8820-1G
2-Cyan-4-(trifluormethyl)phenylboronsäurepinakolester
2-Cyan-4-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₅BF₃NO₂
Molecular Weight: 297,0806 g/mol
MDL: MFCD09878539
CAS-Nummer: 1073355-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR