3-Chlor-5-fluorbenzylamin Hydrochlorid 99%
Lieferant: Thermo Scientific
|
Achtung
|
H51131.06EA
520
EUR
H51131.06
3-Chlor-5-fluorbenzylamin Hydrochlorid 99%
3-Chlor-5-fluorbenzylamin Hydrochlorid
Formel:
C₇H₈Cl₂FN Molecular Weight: 196,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11113045 CAS-Nummer: 90389-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...