3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-8529-1GEA 143 EUR
COBBPN-8529-1G
3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester
3-Chlor-5-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₅BClF₃O₂
Molecular Weight: 306,52 g/mol
MDL: MFCD11856039
CAS-Nummer: 942069-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR