5-Brom-2-fluorbenzylamin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL532762-25GEA
552
EUR
SIAL532762-25G
5-Brom-2-fluorbenzylamin Hydrochlorid, Sigma-Aldrich®
5-Brom-2-fluorbenzylamin Hydrochlorid
Formel:
C₇H₈BrClFN Molecular Weight: 240,50 g/mol |
MDL:
MFCD00143426 CAS-Nummer: 202865-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...