4-(trans-2-Carboxyvinyl)benzeneboronic acid, 96%

H53103.MDEA 20.3 EUR
ALFAH53103.MD ALFAH53103.03
4-(trans-2-Carboxyvinyl)benzeneboronic acid, 96%
Purity: 96%
CAS No.: 159896-15-8
Molecular Formula: C9H9BO4
Formula Weight: 191.98
Melting Point: 222-224°
MDL No.: MFCD01075704
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR