N-Fluorbenzolsulfonimid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL392715-5GEA 74.1 EUR
SIAL392715-5G
N-Fluorbenzolsulfonimid, Sigma-Aldrich®
N-Fluorbenzolsulfonimid
Formel: C₁₂H₁₀FNO₄S₂
Molecular Weight: 315,34
MDL: MFCD00144885
CAS-Nummer: 133745-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR