Benzolsulfonylfluorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL282332-5GEA 146 EUR
SIAL282332-5G
Benzolsulfonylfluorid, Sigma-Aldrich®
Benzolsulfonylfluorid
Formel: C₆H₅FO₂S
Molecular Weight: 160,17
MDL: MFCD00007412
CAS-Nummer: 368-43-4
EINECS: 206-707-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR