2-(Methylsulfinyl)phenol

Lieferant: COMBI-BLOCKS INC.

COBBOT-0400-1GEA 277 EUR
COBBOT-0400-1G COBBOT-0400-5G
2-(Methylsulfinyl)phenol
2-(Methylsulfinyl)phenol
Formel: C₇H₈O₂S
MDL: MFCD00127350
CAS-Nummer: 1074-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR