N-Isopropyl-4-fluor-2-nitroanilin

Lieferant: FLUOROCHEM LTD (CHEMPUR)

CHMPCFF1195.1EA 522 EUR
CHMPCFF1195.1 FLUO017168-2.5G
N-Isopropyl-4-fluor-2-nitroanilin
N-Isopropyl-4-fluor-2-nitroanilin
Formel: C₉H₁₁FN₂O₂
MDL: MFCD00798379
CAS-Nummer: 146366-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR