N-tert-Butyl-4-fluor-2-nitroanilin

CHMPCFF865.EA 0 EUR
CHMPCFF865.
N-tert-Butyl-4-fluor-2-nitroanilin
N-tert-Butyl-4-fluor-2-nitroanilin
Formel: C₁₀H₁₃FN₂O₂
CAS-Nummer: 239080-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR