2-(Difluormethylthio)benzamid

Lieferant: CHEMPUR

CHMPFL5766.5EA 0 EUR
CHMPFL5766.5 CHMPFL5766.10
2-(Difluormethylthio)benzamid
2-(Difluormethylthio)benzamid
Formel: C₈H₇F₂NOS
CAS-Nummer: 261944-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR